Clear bottom, black and white, polystyrene microplates, 384-well, Corning®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3985 3643 3985BC 3540 3542 3544 3766 3762 3764 3768 3770 3764BC 3770BC 3763 3765 3767 3769 4588 4596 4690
734-1200EA 1997.06 EUR InStock
734-1200 734-4118 734-4168 734-4086 734-4087 734-4088 CORN3766 CORN3762 CORN3764 CORN3768 738-0210 CORN3764BC 738-0211 CORN3763 CORN3765 CORN3767 CORN3769 CORN4588 CORN4596 CORN4690
Clear bottom, black and white, polystyrene microplates, 384-well, Corning®
Mikrotiterplatten HTS-Mikrotiterplatten
PS, flat bottom. Suitable for fluorescent and luminescent assays using either top or bottom detection plate readers.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge