Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Volumetric flasks, class B, with tapered stopper, Corning®

Lieferant: Corning

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

5641P-100 5641P-1L 5641P-50 5641P-500 5641P-10 5641P-250
CORN5641P-100EA 168.12 EUR InStock
CORN5641P-100 CORN5641P-1L CORN5641P-50 CORN5641P-500 CORN5641P-10 CORN5641P-250
Volumetric flasks, class B, with tapered stopper, Corning®
Kolben Messkolben
Made of PP, translucent, with tapered PP stopper.

  • With an individually calibrated graduation line
  • Recommended routine cleaning up to 60 °C will preserve graduations
  • Exposure to temperatures above 60 °C may affect accuracy

Reusable plastic flasks are manufactured to Class B tolerances as defined by ISO 1042.

The accuracy of volumetric ware depends on the care used in calibrating it, using the correct type of ware for the application, handling the ware properly and ensuring the ware is clean. Calibration of volumetric ware is usually done at 20 °C and the ware should be used at approximately this temperature. Refrigerated liquids should be allowed to come to room temperature before measuring them. Under normal use and care, the calibration of volumetric ware will not change. Do not expose volumetric ware to excessive heat, defined as temperatures approaching the Corning-defined upper service limit. The polymers from which the Corning line is manufactured have unique thermal characteristics. The polymer temperature capability is described above.

It is important to recognise the difference in specifications regarding DIN and ASTM standards. For reference, ASTM Class A and B specifications are more precise than DIN Class A and B specifications, respectively.

Height dimension listed is for flask only, without stopper.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge