Funnels, with short stem, Corning®
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
6120P-50 6120P-40 6120P-30 6120P-150 6120P-120 6120P-75 6120P-100
CORN6120P-50EA 31.13 EUR InStock
CORN6120P-50 CORN6120P-40 CORN6120P-30 CORN6120P-150 CORN6120P-120 CORN6120P-75 CORN6120P-100
Funnels, with short stem, Corning®
Trichter Filtertrichter Filtertrichter
Funnels, made of durable PP, translucent.
  • Very good chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Allow safe and easy transfers of solids or liquids from one vessel to another.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge