Magnetic stirring bars, plain, PTFE
Lieferant: COWIE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
001.212 001.215.6 001.230 001.225.8 001.250.7 001.235.7 001.250 001.260
COWI001.212EA 1.8 EUR InStock
COWI001.212 COWI001.215.6 COWI001.230 COWI001.225.8 COWI001.250.7 COWI001.235.7 COWI001.250 COWI001.260
Magnetic stirring bars, plain, PTFE
Rührer Magnetrührstäbchen
Plain PTFE magnetic stirring bars have similar performance to cylindrical bars, but provide more turbulence at low speeds.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge