Magnetic stirring bars, PTFE/rare earth, TURBO™ flute
Lieferant: COWIE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
001.3450.RE
COWI001.3450.REEA 142 EUR InStock
COWI001.3450.RE
Magnetic stirring bars, PTFE/rare earth, TURBO™ flute
Rührer Magnetrührstäbchen
These stirring bars are made of extra power samarium-cobalt magnets which are chemically inert and have increased magnetic strength compared to Alnico.
  • Totally resistant to demagnetisation
  • Multi-faceted stirrer for general use
Temperature resistance: −200 to +280 °C.
Temperature resistance: −200 to +280 °C.
Achtung! Bitte beachten: The high magnetic coupling power of rare earth magnets used in these stirring bars can give rise to erratic stirring effects such as magnet migration and tumbling while the powerful attraction between the stirrer and the drive magnet system can give rise to the formation of PTFE particles caused by abrasion between stirrer magnet and the container base.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge