Magnetic stirring bar retrievers, flexible, PTFE
Lieferant: COWIE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
004.330F
COWI004.330FEA 102 EUR InStock
COWI004.330F
Magnetic stirring bar retrievers, flexible, PTFE
Rührer Rührwerkzeuge
Retrivers are used for the removal of stirrer magnets from vessels of all kinds.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge