Tubing connectors, L-shaped, Bayonet type
Lieferant: COWIE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
016.1005.2 016.1002.2 016.1002.5.2 016.1003.2 016.1003.7.2 016.1004.9.2 016.1004.2
COWI016.1005.2EA 27.5 EUR InStock
COWI016.1005.2 COWI016.1002.2 COWI016.1002.5.2 COWI016.1003.2 COWI016.1003.7.2 COWI016.1004.9.2 COWI016.1004.2
Tubing connectors, L-shaped, Bayonet type
Fittings und Verbinder
L-shaped right angle connectors, made of PTFE, for tubings.
  • Very good chemical resistance
  • Very good temperature resistance
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge