Joint clamps for standard ground joints, Cowie®
Lieferant: COWIE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
030.045.1 030.012.1 030.010.1
COWI030.045.1EA 14.1 EUR InStock
COWI030.045.1 COWI030.012.1 COWI030.010.1
Joint clamps for standard ground joints, Cowie®
Klemmen Schliffklemmen
These PTFE joint clamps with totally encapsulated steel springs can be adjusted by hand and fit standard glass joints.
  • Chemically inert and totally unaffected by laboratory chemicals
  • Temperature resistant and maintain strength right up to 280 °C
  • Can be simply adjusted by compression-extension by hand
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge