Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Shaft guides, High Performance (HP), for standard taper ground glass joints

Lieferant: COWIE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

005.00.0619 005.00.0624 005.00.0819 005.00.0824 005.00.1024 005.00.1029 005.00.1045 005.00.1229 005.00.1634 005.00.1645 005.00.1945 005.00.0824.PK 005.00.1024.PK 005.00.1029.PK 005.00.1045.PK 005.00.1229.PK 005.00.1634.PK 005.00.1645.PK 005.00.1945.PK 005.00.0819.PK PC710.0819.PK PC710.0825.PK PC710.1019.PK PC710.1025.PK PC710.1225.PK PC710.1625.PK PC710.1925.PK 005.0.1029 005.0.1945.PK 005.0.1945 005.0.1634.PK 005.0.1634 005.0.1229.PK 005.0.1229 005.0.1045.PK 005.0.1045 005.0.1034.PK 005.0.1029.PK 005.0.1024.PK 005.0.1024 005.0.0824.PK 005.0.0824 005.0.0819.PK 005.0.0819 005.0.0624 005.0.0619 005.0.1034
COWI005.00.0619EA 182 EUR InStock
COWI005.00.0619 COWI005.00.0624 COWI005.00.0819 COWI005.00.0824 COWI005.00.1024 COWI005.00.1029 COWI005.00.1045 COWI005.00.1229 COWI005.00.1634 COWI005.00.1645 COWI005.00.1945 COWI005.00.0824.PK COWI005.00.1024.PK COWI005.00.1029.PK COWI005.00.1045.PK COWI005.00.1229.PK COWI005.00.1634.PK COWI005.00.1645.PK COWI005.00.1945.PK COWI005.00.0819.PK COWIPC710.0819.PK COWIPC710.0825.PK COWIPC710.1019.PK COWIPC710.1025.PK COWIPC710.1225.PK COWIPC710.1625.PK COWIPC710.1925.PK COWI005.0.1029 COWI005.0.1945.PK COWI005.0.1945 COWI005.0.1634.PK COWI005.0.1634 COWI005.0.1229.PK COWI005.0.1229 COWI005.0.1045.PK COWI005.0.1045 COWI005.0.1034.PK COWI005.0.1029.PK COWI005.0.1024.PK COWI005.0.1024 COWI005.0.0824.PK COWI005.0.0824 COWI005.0.0819.PK COWI005.0.0819 COWI005.0.0624 COWI005.0.0619 COWI005.0.1034
Shaft guides, High Performance (HP), for standard taper ground glass joints
Rührer Rührschäfte
High Performance (HP) shaft guides provide an effective guide for use with glass and metal shaft stirrers over a wide range of temperatures without shedding particles from a specially formulated PTFE-PEEK seal whilst maintaining vacuum. The moulded-in PTFE-PEEK guide ring located at the end of the taper provides a guide to assist alignment of the shaft.

Shaft guides have an NPT thread for direct assembly into PTFE reactor lids or standard tapers for use with glass joints.

Achtung! Bitte beachten: PEEK has reduced chemical resistance compared to PTFE and may be affected by acids, phenols and halogen based compounds.

Maximum recommended speeds are 500 rpm continuous or 800 rpm intermittent.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge