MPX series couplings, medical grade
Lieferant: CPC COLDER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MPX17839B25 MPX22639MB25 MPX22803MB25 MPX22839MB25 MPX32003 MPX32003B25 MPX32039 MPX32039B25 MPXK17839B25
CPCCMPX17839B25EA 495 EUR InStock
CPCCMPX17839B25 CPCCMPX22639MB25 CPCCMPX22803MB25 CPCCMPX22839MB25 CPCCMPX32003 CPCCMPX32003B25 CPCCMPX32039 CPCCMPX32039B25 CPCCMPXK17839B25
MPX series couplings, medical grade
Fittings und Verbinder
MPX Series couplings add ease of use and security to your most critical fluid handling applications.
  • Operating temperature: −40 to 149 °C
  • Easy to operate
  • BSE/TSE free materials
  • Parting line free hose barb
  • Ergonomic thumb latch
  • Sterilisable by autoclave, Et0, e-beam or gamma
These couplings provide optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
Zertifizierungen: USP Class VI, ADCF certified and meets biocompatibility requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge