Steam-Thru II configuration couplings
Lieferant: CPC COLDER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
STC2021000
CPCCSTC2021000EA 1410 EUR InStock
CPCCSTC2021000
Steam-Thru II configuration couplings
Fittings und Verbinder
Steam-Thru II Connections offer the flexibility of 'steam on' and 'steam off' functionality. The innovative design allows the valve to be returned to the steam position enabling a second SIP cycle following media transfer.
Materials of construction include USP class 6 animal free (BSE/TSE) polysulphone, USP class 6 andimal free (BSE/TSE) silicone O-rings and seals.
Zertifizierungen: USP Class VI, BSE/TSE free materials (ADCF).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge