Hook2Hook coil tether with swivel, DBI-SALA®, 3M™
Lieferant: CAPITAL SAFETY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1500159
CPSD1500159EA 0 EUR InStock
CPSD1500159
Hook2Hook coil tether with swivel, DBI-SALA®, 3M™
Fallschutz Verbindungsmittel und Seile
DBI-SALA® Hook2Hook coil tether tool lanyard, coil tether features a swivel hook, making tools like screwdrivers and wrenches easier to use. It is a tethering solution specifically designed for tools weighing up to 2,3 kg. With a relaxed length of 4", and a stretched length of 62", this tool lanyard stays out of the way when not in use. An extra-thick PU coating around the coil prevents kinking and allows the coil to retain its form.
  • Compact design stays close to the body
  • Secure tools up to 2,3 kg
  • Lightweight dual locking aluminum carabiners
  • Vinyl-coated braided steel coil
  • Resistant to heat and to sharp objects
Suitable for use from holsters, belts, harnesses and tool pouches alike, the Hook2Hook Coil Tether is a versatile solution that is both resistant to heat and to sharp objects. Lightweight aluminum carabiners on either side of the extension feature locking screw-down nuts that cannot be removed.
Capacity: 2,3 kg
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge