Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Retrieval self-retracting lifeline, Sealed-Block™ DBI-SALA, 3M™

Lieferant: CAPITAL SAFETY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

3400856 3400881
CPSD3400856EA 0 EUR InStock
CPSD3400856 CPSD3400881
Retrieval self-retracting lifeline, Sealed-Block™ DBI-SALA, 3M™
Fallschutz Höhensicherungsgeräte und Fallbegrenzer
Sealed-Blok™ family has the most comprehensive line of sealed SRLs in the industry and decades of proven field service in the toughest environments.

  • Pivoting anchorage loop
  • Aluminum housing and stainless steel life line
  • 2267 kg Rated handle
  • Impact indicator
  • Tripod / davit arm bracket option
  • 5 mm Ø rope and 1633 kg gate
  • RSQ options and FAST-Line lifeline assembly

The heavy duty sealed design keeps the critical working components free of dirt, grease, water and chemicals. The range also includes a dual range SRL and automatic self-rescue device. Ideal for use in harsh environments such as offshore, confined space, salt water and dirty environments.

Zertifizierungen: Meets IP68 rating, EN 360, EN1496 clB.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge