Aluminium tripod, DBI-SALA, 3M™
Lieferant: CAPITAL SAFETY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8563158 8563159
CPSD8563158EA 0 EUR InStock
CPSD8563158 CPSD8563159
Aluminium tripod, DBI-SALA, 3M™
Fallschutz Höhensicherungsgeräte und Fallbegrenzer
Tripod with adjustable locking legs, safety chains, top pulley and winch bracket.
  • Available in two heights
Weight capacity: 205 kg
Zertifizierungen: EN 795 Class-B
Bestellinformation: Winch and SRL are not included.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge