Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Silberdiethyldithiocarbamat, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

D3132-25G D3132-5G
D3132-25GEA 281 EUR InStock
D3132-25G D3132-5G
Silberdiethyldithiocarbamat, Sigma-Aldrich®
Silberdiethyldithiocarbamat
Spin trap used in conjunction with Fe²⁺ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO₂ has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes. Also used for colorimetric determination of small quantities of arsenic. Silver diethyldithiocarbamate has been used in cell culture. Also used for colorimetric determination of small quantities of arsenic. Spin trap used in conjunction with Fe²⁺ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO₂ has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

  • Inhibits induction of macrophage nitric oxide synthase
  • Silver diethyldithiocarbamate (AgDDC) is used to estimate the level of arsenic in human
  • Inhibits induction of macrophage nitric oxide synthase


Formel: (C₂H₅)₂NCSSAg
Melting Pt: 172…175 °C (Lit.)
Storage Temperature: Kühlschrank
MDL Number: MFCD00004929
CAS-Nummer: 1470-61-7
EINECS: 216-003-7
UN: 3077
ADR: 9,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c07412a6ac577902b87e980dc94c68b5023241ea&vid=c07412a6ac577902b87e980dc94c68b5023241ea&prid=web&clid=SAPDAM
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge