Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

4-(Dimethylamino)zimtaldehyd chromogenes Reagenz für Indole und Flavanole, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

M-Clarity™ Qualitätsstufe = MQ200

D4506-10G D4506-1G D4506-25G D4506-5G
D4506-10GEA 210 EUR InStock
D4506-10G D4506-1G D4506-25G D4506-5G
4-(Dimethylamino)zimtaldehyd chromogenes Reagenz für Indole und Flavanole, Sigma-Aldrich®
4-(Dimethylamino)zimtaldehyd
Reagent for assay of indole product of apotryptophanase and tryptophanase. DMAC produces colored adducts with flavanols for subsequent HPLC. It has been used in a chromogenic method for quantifying proanthocyandines in cranberry powder.

  • 4-(Dimethylamino)cinnamaldehyde (DMAC) has been used in the serine protease inhibition assay
  • It has been used for the detection of (epi)-flavan-3-ols and proanthocyanidin polymers in Brassica napus seeds
  • It has been used for the determination of phenolic compounds in leaf samples
  • It has been used for the staining of flavan-3-ols in plant samples


Formel: C₁₁H₁₃NO
Melting Pt: 138,..140 °C (Lit.)
Storage Temperature: Gefrierschrank
MDL Number: MFCD00007002
CAS-Nummer: 6203-18-5
EINECS: 228-267-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=964b8d0ff27a466c1bd1e47f2124d3da6cfee415&vid=964b8d0ff27a466c1bd1e47f2124d3da6cfee415&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (colour) Yellow to very dark yellow and orange to very dark orange
Appearance (form) Powder
Purity (HPLC area %) ≥97.5 %
Solubility (colour) Yellow to very dark yellow to very dark red
Solubility (turbidity) Clear to hazy
Solubility (method) 250 mg in 5 ml chloroform/ethanol (1:1)
Proton nmr spectrum Conforms to structure
Recommended retest period 48 months

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge