Packaging for diagnostic specimen, Polymed
Lieferant: DAKLAPACK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
15004
DAKL15004EA 686 EUR InStock
DAKL15004
Packaging for diagnostic specimen, Polymed
Versandtaschen und Transportbehälter
Ideal for sending diagnostic specimens.
  • Constructed from a strong PVC vinyl which can be used hundreds of times
  • Easily opened and closed via the easy to use zip closure
On the front of the Polymed there will be a clearly visible transparent window. Insert double sided address card in to the window and pop the Polymed into the post box. The Polymed system is suitable for sending via the post box, saving time, money and most of the packaging can be reused.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge