Mailing boxes, mini
Lieferant: DAKLAPACK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
20121
DAKL20121EA 95.3 EUR InStock
DAKL20121
Mailing boxes, mini
Versandtaschen und Transportbehälter
Mini mailing boxes are designed for sending diagnostic samples of Category B.
  • Cardboard material printed with UN-number and logo as per regulations
Zertifizierungen: Approved (BAM and postal organisations) as external packaging for diagnostisc samples of Category B, as long as used in accordance with packaging instruction P650 in combination with safetybag and absorbent tube holders.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge