Face shield, Turboshield™
Lieferant: HONEYWELL SAFETY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1031743 1031744 1031747
DALL1031743EA 27.6 EUR InStock
DALL1031743 DALL1031744 DALL1031747
Face shield, Turboshield™
Gesichtsschutz
The Turboshield™ is a great value faceshield that offers easy visor exchange, is more comfortable to wear and offers better protection.
  • Simple push‐button release system allows for easy visor exchange in seconds
  • Breathable, removable, washable headband
  • Toric lens provides excellent optics, increased field of view and added chin coverage without bulky attachments
  • Ergonomic, easy‐to‐use adjustment knobs
Applications include grinding, welding, chemical, construction, food services, manufacturing, medical, oil and gas, pharmaceutical, steel and metals.
Zertifizierungen: AS/NZS 1337.1:2010
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge