Earplugs dispenser, Leight® Source 500
Lieferant: HONEYWELL SAFETY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3301272 3301273 3301271 3301165
DALL3301272EA 130 EUR InStock
DALL3301272 DALL3301273 DALL3301271 DALL3301165
Earplugs dispenser, Leight® Source 500
Spender für PSA
The Leight Source 500 dispenser provides a highly visible permanent hearing protection source.
  • Heavy-duty anodized aluminum allowing constant use
  • Wall mounting for a user-friendly access
  • Crank handle to dispense earplugs
Suitable for large factories and process industries with Max®, Max Lite®, Laser Lite®, Multi Max®, Matrix™, FirmFit™, Quiet® earplugs.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge