Heat resistant welding gloves, GFA115K
Lieferant: DELTA PLUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
GFA115K
DELPGFA115KEA 191 EUR InStock
DELPGFA115K
Heat resistant welding gloves, GFA115K
Handschuhe
These lambskin full grain leather gloves with Kevlar® threads provide excellent mechanical protection and restricted heat protection.
  • Town cut, added-on thumb
  • Good dexterity
  • Long cuff provides protection for forearm
Suitable for welding applications, metal work and industrial maintenance.
Zertifizierungen: EN 388:2016 (2111X), EN 407 (41XX4X), EN 12477:2001/A1:2005
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge