Filter Bags for Homogeniser
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
15005
DELT15005EA 1200 EUR InStock
DELT15005
Filter Bags for Homogeniser
Beutel Homogenisationsbeutel
These are sterile bags having a full size filter dividing them into two identical pouches, in which sample is inserted into one compartment. After sample process, solid particles remain in their original compartment while liquid will be filtered into the other compartment, thus allowing a convenient liquid pipetting.
  • Beta irradiation
  • Graduated for marking
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge