Autoclave Bags
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
200100 200311 200312 200310
DELT200100EA 105 EUR InStock
DELT200100 DELT200311 DELT200312 DELT200310
Autoclave Bags
Beutel Entsorgungsbeutel
Autoclave resistant bags, printed with the biohazard graphic symbol and instructions of use in five languages.
Achtung! Bitte beachten: Do not close the bags hermetically when placing them in the autoclave at 121 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge