Drums, stackable, with tamper evident cap
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
206025
DELT206025EA 28.6 EUR InStock
DELT206025
Drums, stackable, with tamper evident cap
Fässer, Fässer, Trommeln
Stackable drums, made of HDPE, natural.
  • Hermetical closure
  • For alimentary use
They embody a handle and a tamper evident, ribbed screw red cap, made of HDPE, with internal joint and internal security ring.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge