Centrifuge tubes, with screw caps, EUROTUBO®
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
401402 301403 301402 401403
DELT401402EA 734 EUR InStock
DELT401402 DELT301403 DELT301402 DELT401403
Centrifuge tubes, with screw caps, EUROTUBO®
Röhrchen Zentrifugenröhrchen
Made of PP, translucent or PS, clear, with green HDPE screw cap.
  • PP: autoclavable
  • PP: Withstand forces up to 15000×g
  • PS: Withstand forces up to 7500×g
The external skirt allows the tubes to remain free-standing. Supplied screwed. The sterile model (ethylene oxide) is supplied individually in flow-pack bag, with indication of batch, expiration date, etc. according to the guidelines for sterile products.
Achtung! Bitte beachten: For autoclaving, the cap should be loose on the thread and not tightly fitted.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge