Reaktionsgefäße
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4092.6N
DELT4092.6NEA 804 EUR InStock
DELT4092.6N
Reaktionsgefäße
Röhrchen Reaktionsgefäße
Made of ultra clear polypropylene.
  • Autoclavable
  • Certified RNAse and DNAse free
  • Feature a frosted writing surface on side and on closure for better sample identification
  • Lids are easily pierceable
  • Tubes have moulded-in graduations in 100 μl increments up to 2 ml
  • Can be used at temperatures from –90 up to 121 °C
  • Withstand centrifugation up to 17000×g
Easy handling: tubes can be opened and closed with one hand.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge