Strips of 8 PCR tubes with attached flat cap
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4096.2N
DELT4096.2NEA 623 EUR InStock
DELT4096.2N
Strips of 8 PCR tubes with attached flat cap
Röhrchen PCR-Gefäße
Made of PP.
  • RNAse, DNAse and pyrogen free
  • Excellent chemical resistance
  • Excellent temperature resistance
Every cap embodies a shield in order to prevent contamination when being opened. Positive sealing will fully protect the contents from evaporation during the whole thermal cycle. These strips can be cut to any length while each tube has its own attached cap.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge