Wash bottles, narrow neck
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
44310000 44300000
DELT44310000EA 259 EUR InStock
DELT44310000 DELT44300000
Wash bottles, narrow neck
Flaschen Spritzflaschen
HDPE, translucent, with blue HDPE screw cap, delivery tube and seal cap, natural.
  • High resistance to all liquids, acids and bases
  • Wide neck for safe and easy filling
  • Caps ribbed for a better handling
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge