Screw caps for tubes
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
617100 617200 617300 617000
DELT617100EA 344 EUR InStock
DELT617100 DELT617200 DELT617300 DELT617000
Screw caps for tubes
Verschlüsse Verschlüsse für Reagenz- und Probenröhrchen
Screw caps made of polypropylene and aluminium with an internal liner made of rubber.
  • Both models of caps are fully autoclavable and are pharmaceutical quality
  • Excellent chemical resistance
  • Excellent temperature resistance
Screw caps for round bottom borosilicate glass tubes and flat bottom soda glass tubes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge