Drucken
Transport containers
Lieferant: DELTALAB
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
900028 979930 900025
631-9780EA 17.9 EUR
631-9780 DELT979930 DELT900025
Transport containers
Objektträger-Versandbehälter
Designed for slide transport and protection.
  • Made of polypropylene
  • Easy snap-lock
  • Not indicated for liquid samples
Compartment shape ensures tight fit: no risk of damaging slides and to send by mail, recommend to use special envelopes.