Screw thread vials
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
900220-928000 900211 900205 900220 900225
DELT900220-928000EA 0 EUR InStock
DELT900220-928000 DELT900211 DELT900205 DELT900220 DELT900225
Screw thread vials
Vials
These flat bottom screw thread vials are made of ultra clear borosilicate glass.
  • Fully autoclavable
Bestellinformation: Caps are supplied separately.
Verpackung: Supplied in trays individually shrink-wrapped.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge