Screw caps for screw neck vials
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
918000 922000 928000
DELT918000EA 670 EUR InStock
DELT918000 DELT922000 DELT928000
Screw caps for screw neck vials
Verschlüsse Chromatographieampullen-Verschlüsse
These screw caps are made of autoclavable PP.
  • Internal silicone joint of pharmaceutical quality
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge