Microtube racks, incubation, round
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M-026 M-024
DELTM-026EA 231 EUR InStock
DELTM-026 DELTM-024
Microtube racks, incubation, round
Gestelle Reagenzglasgestelle
PP
Even when the rack is fully loaded, tubes are completely immersed while tops rest above the rack, floating in a beaker of water. Perfect for use in benchtop incubation. The rack is easily unloaded by lightly pressing it down against the bench until the legs touch, releasing the tubes. Racks are 6,4 mm thick with 19 mm long legs and a central handle.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge