Tube racks, universal
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M-522 M-520 M-523
DELTM-522EA 183 EUR InStock
DELTM-522 DELTM-520 DELTM-523
Tube racks, universal
Gestelle Reagenzglasgestelle
PP, linkable to other racks.
  • Versatile rack with different capacities on each of the four sides
  • Alpha-numeric matrix on the four sides for better sample location
  • Suitable for freezers and autoclaves
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge