Microtube racks, reversible, with removable handles
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M-563A
DELTM-563AEA 204 EUR InStock
DELTM-563A
Microtube racks, reversible, with removable handles
Gestelle Reagenzglasgestelle
PP, reversible.
  • Stackable
  • By removing the handles, units can be firmly anchored laterally to one another
The racks have two sides for different tubes: one side holds up to 80 tubes of 12 mm diameter (for example 10×75 mm or 12×75 mm tubes, microtubes, etc.) and the other side holds up to 60 microtubes, PCR, etc.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge