Test tube racks
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
U-15128A
DELTU-15128AEA 52 EUR InStock
DELTU-15128A
Test tube racks
Gestelle Reagenzglasgestelle
Test tube racks, made of PP, with transparent lid.
  • Excellent chemical resistance
  • Resist –196 °C and autoclavable up to 121 °C
  • Eays to clean
Available in fluorescent colours with transparent lid to help identification. Sections will hold 25 tubes up to 17 mm diameter and 125 mm height.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge