Conductive Tote Boxes
Lieferant: DESCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
239305 239345
DESC239305EA 26.7 EUR InStock
DESC239305 DESC239345
Conductive Tote Boxes
Boxen ESD-Sicherheitsboxen
Conductive tote boxes are made from injection moulded conductive polypropylene.
  • Flat base, stackable
  • Rv: 1×10³ to <1×10⁶ Ω
  • Custom-made dividers are available for PCB containers
  • Machined from approximately 2 mm thick conductive plastic or Cortronic® B-flute static dissipative cardboard
  • Many shapes and sizes possible
  • Process is suited for small and medium quantities
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge