BioTherm Dry Ice Systems (Category B Compliant)
Lieferant: DGP INTELSIUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BT104 BT111
DGINBT104EA 63.7 EUR InStock
DGINBT104 DGINBT111
BioTherm Dry Ice Systems (Category B Compliant)
Versandtaschen und Transportbehälter
The BioTherm Dry Ice range is designed forthe safe transportation of deep frozen samples to IATA, ADR and DOT (49 CFR) transportregulations. The systems offer a variety of options for the safe transit of Category B shipments, as complete systems or as overpacks.
  • Infectious samples
  • Diagnostic samples
  • Clinical trials
  • Screening programmes
  • Eradication schemes
Category B systems provided with compliant transport bags and absorbent.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge