Alarm thermometer, MM125
Lieferant: DICKSON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MM125
DICKMM125EA 191 EUR InStock
DICKMM125
Alarm thermometer, MM125
Thermometer Digitalthermometer
MM125 is a simple, cost effective temperature monitoring tool which is used as a backup to a traditional data logger in vaccine or drug monitoring application.
  • Tamper-resistant audible/visual alarms
  • Two thermistor probes submersed in glass beads for stabilized readings
  • Tracks min/max temperatures
  • Easy to read display
Lieferumfang: The MM125 supplied with the temperature sensor submersed in glass beads instead of a glycol solution (glass beads and glycol are recommended by the CDC for vaccine (including VFC) storage.)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge