Prisms
Lieferant: DIDALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
POD010615
DIDAPOD010615EA 17.5 EUR InStock
DIDAPOD010615
Prisms
Lehrmittel Physik-Lehrmittel Lehrmittel für die Optik
Prism made of standard optical glass with index near to 1,52. Equilateral dispersing prisms are used for wavelength separation applications. Intended for demonstration experiments of light dispersion.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge