Diode laser, green, Class II and IIIa
Lieferant: DIDALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
POD013132
DIDAPOD013132EA 342 EUR InStock
DIDAPOD013132
Diode laser, green, Class II and IIIa
Lehrmittel Physik-Lehrmittel Lehrmittel für die Optik
Extremely luminous beam, ideal for classroom experiments. Class IIIa (5 mW Power)
  • Highly miniaturised system with glass optic components
Lieferumfang: Provided with Ø 10 mm rod, mains power supply & adapter for use in Class II.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge