Laurent polarimeters
Lieferant: DIDALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
POD068570 POD068570_UK
DIDAPOD068570EA 1030 EUR InStock
DIDAPOD068570 DIDAPOD068570_UK
Laurent polarimeters
Polarimeter
The Laurent polarimeter is composed of one lens fitted on graduated rule, and one housing for observation tubes, within which the substances to be analysed are introduced.
Designed for measurement of the polarisation direction in light through an optical medium.
Lieferumfang: Supplied with two observation tubes (100 and 200 mm)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge