Standard prismatic optical bench, DidaFirst
Lieferant: DIDALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
POF010122 POF010124 POF010125 POF010126 POF010112 POF010110
DIDAPOF010122EA 175 EUR InStock
DIDAPOF010122 DIDAPOF010124 DIDAPOF010125 DIDAPOF010126 DIDAPOF010112 DIDAPOF010110
Standard prismatic optical bench, DidaFirst
Lehrmittel Physik-Lehrmittel Lehrmittel für die Optik
Economical, recent design of prismatic optical bench
  • Anodised aluminium
  • Graduated
  • Adjustable feet available
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge