Microtube membranes, breathable, Breathe-EASIER
Lieferant: DIVERSIFIED BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BMTM-2000
DIVEBMTM-2000EA 114 EUR InStock
DIVEBMTM-2000
Microtube membranes, breathable, Breathe-EASIER
Versiegeln Verschlussfolien
Breathe-EASIER micropermeable membranes are ideal for cell growth, concentrating samples, and gas exchange. Formatted with an easy to use finger tab, membranes are precut to fit a variety of tube sizes. Membranes are gamma irradiated to ensure sterility and are free of cytotoxins.
  • Microtube cap closures will not bind in centrifuge rotors
  • Convenient scoring makes liner removal quick and easy
  • White non woven rayon membrane rapidly transmits water vapour for accelerated growth
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge