Colour-coded Tough-Tags® labels
Lieferant: DIVERSIFIED BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
COLB-1000 COLR-1000 COLG-1000 COLY-1000 COLW-1000
DIVECOLB-1000EA 144 EUR InStock
DIVECOLB-1000 DIVECOLR-1000 DIVECOLG-1000 DIVECOLY-1000 DIVECOLW-1000
Colour-coded Tough-Tags® labels
Etiketten
These Tough-Tags® polyester colour-coded labels make it easy to differentiate samples and identify what you need.
  • Temperature resistant from −80 to +90 °C
  • Translucent writing area keeps contents visible
  • Set consists of a colour-coded rectangular label and spot
The labels are repositionable for up to thirty minutes as the adhesive strengthens over time.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge