Thermal transfer Cryo-Tags® labels
Lieferant: DIVERSIFIED BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CRTH-2000
DIVECRTH-2000EA 557 EUR InStock
DIVECRTH-2000
Thermal transfer Cryo-Tags® labels
Etiketten
These Cryo-Tags® polyester labels work with most thermal transfer printers to print popular barcode formats for fast and easy sample identification.
  • Temperature resistant from −196 to +80 °C
  • Freezable in liquid and vapour phase nitrogen
  • Solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
  • Resists degradation from scuffing, chemicals, moisture and temperature fluctuations
Print crisp, clear, professional quality labels.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge