WetGrip labels
Lieferant: DIVERSIFIED BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
GRDT-2000 GRDT-1000 GRDT-3000 GRDT-4000 GRDT-6000 GRLA-2016 GRLA-1000 GRLA-2380 GRLA-1050
DIVEGRDT-2000EA 157 EUR InStock
DIVEGRDT-2000 DIVEGRDT-1000 DIVEGRDT-3000 DIVEGRDT-4000 DIVEGRDT-6000 DIVEGRLA-2016 DIVEGRLA-1000 DIVEGRLA-2380 DIVEGRLA-1050
WetGrip labels
Etiketten
These WetGrip labels grip to moist, frosty or frozen surfaces.
  • Temperature resistant to −80 °C
  • Compatible with centrifuge rotors
  • Resistant to chemicals and solvents
  • Adheres to plastics, glass, metals and even paperboard boxes
Available in white laser sheets, white direct thermal rolls, or white hand writable rolls.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge