Labels for petri dishes, PetriStickers™
Lieferant: DIVERSIFIED BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PSTK-1000 PSTK-1050 PSTK-1070 PSTK-1100 PSTK-2000 PSTK-2010 PSTK-2020 PSTK-2030
DIVEPSTK-1000EA 82.9 EUR InStock
DIVEPSTK-1000 DIVEPSTK-1050 DIVEPSTK-1070 DIVEPSTK-1100 DIVEPSTK-2000 DIVEPSTK-2010 DIVEPSTK-2020 DIVEPSTK-2030
Labels for petri dishes, PetriStickers™
Etiketten
PetriStickers™ allow for petri dishes to be labeled quickly, neatly and reliably.
  • Temperature resistant from +4 to +42° C
  • Water-resistant ink
Simply remove the label's protective backing and apply it to the back of a petri dish. After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge