Radtape™ labels
Lieferant: DIVERSIFIED BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
RAD-10
DIVERAD-10EA 259 EUR InStock
DIVERAD-10
Radtape™ labels
Etiketten
These RadTape™ labels are used to identify autograms. They are phosphorescent, non-radioactive adhesive labels that may be cut to the specific size and shape needed for each individual membrane, dried gel or microarray.
  • Ideal for chemilluminescence and radiological autography
  • Great for documentation and archiving
  • Use your own codes and symbols
  • Fast and easy marking system
An ordinary pen may be used to mark the label using the investigator’s own codes and symbols. The custom label may then be peeled from its backing and affixed to the membrane, gel or microarray. Once placed against film, Radtape™ exposes a negative image of the markings directly onto the autorad, thus producing a permanent record of the experiment.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge