Station for Tough-Tags
Lieferant: DIVERSIFIED BIOTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TTSA-9000
DIVETTSA-9000EA 202 EUR InStock
DIVETTSA-9000
Station for Tough-Tags
Etikettenhalter
Tough tag station holds up to six rolls of Tough-tags®, Tough-spots® and Cryo-tags® labels. The ledge in the station is used to write on labels and the straight edge is to tear off the labels.
  • Built-in cutting edge
  • Convenient writing surface
  • Holds multiple rolls for easy transport
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge